Learn More
2-Phenoxynicotinic acid, 98%, BLD Pharm™
Product Code. 30968289
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30968289 25 g 25g
30956620 100 g 100g
30968189 1 g 1g
30949229 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30968289 Supplier BLD Pharm Supplier No. BD1447925g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 35620-71-4
Molecular Formula C12H9NO3
MDL Number MFCD00014629
Quantity 25 g
SMILES OC(=O)C1=CC=CN=C1OC1=CC=CC=C1
Molecular Weight (g/mol) 215.20
Percent Purity 98%
Chemical Name or Material 2-Phenoxynicotinic acid