Learn More
2-[(Phenylsulfanyl)methyl]benzenecarboxylic Acid, TRC
Product Code. 30340694
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30340694 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30340694 Supplier Toronto Research Chemicals Supplier No. TRCB481480500MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Especificaciones
Chemical Name or Material 2-[(Phenylsulfanyl)methyl]benzenecarboxylic Acid
CAS 1699-03-2
Molecular Formula C14H12O2S
Quantity 500 mg
Synonym o-Toluic acid, alpha-(phenylthio)- (6CI,7CI,8CI), 2-[(Phenylthio)methyl]benzoic acid, 2-(Phenylthiomethyl)benzoic acid, 2-[(Phenylsulfanyl)methyl]benzoic acid, alpha-(Phenylthio)-o-toluic acid
InChI Formula InChI=1S/C14H12O2S/c15-14(16)13-9-5-4-6-11(13)10-17-12-7-2-1-3-8-12/h1-9H,10H2,(H,15,16)
SMILES OC(=O)c1ccccc1CSc2ccccc2
IUPAC Name 2-(phenylsulfanylmethyl)benzoic acid
Molecular Weight (g/mol) 244.31
Formula Weight 244.0558
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.