Learn More
2-(Trifluoromethoxy)benzenesulfonamide, 98%, BLD Pharm™
Product Code. 30986987
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
Unit Size:
500g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30986987 1 g -
30982029 10 g -
30975707 5 g 5g
30970463 100 g -
30968920 500 g 500g
30977823 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30986987 Supplier BLD Pharm Supplier No. BD104501g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 37526-59-3
Molecular Formula C7H6F3NO3S
MDL Number MFCD01320751
Quantity 1 g
SMILES O=S(C1=CC=CC=C1OC(F)(F)F)(N)=O
Molecular Weight (g/mol) 241.19
Percent Purity 98%
Chemical Name or Material 2-(Trifluoromethoxy)benzenesulfonamide