Learn More
Brannan™ 200 Series Specific Gravity Hydrometers
Product Code. 15949330
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
15949330 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15949330 Supplier Brannan™ Supplier No. 10/500/0
  • ONLINE EXCLUSIVE PRICE
    26.11€
    15.52€ / 1 pièces
    Économisez 10.59€
    40% de réduction
Expédition estimée: 08-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Compares the relative density of a given liquid with that of water at 15.6°C (60°F). Brannan™ 200 Series Specific Gravity Hydrometers are manufactured from soda glass, steel shot ballast and synthetic wax resin.
  • 250mm in length
  • Scale subdivision 0.002
  • Accuracy of ± 0.002 specific gravity
  • Includes protective plastic tube
  • Available adjusted for either low or medium surface tension
  • Available for numerous specific gravity ranges
Specifications
Type 200 Series Specific Gravity Hydrometers
Description ISO 650, 916
Material Glass
For Use With (Application) Low surface tension