Learn More
BRAND™ 24-well Insert 2in1, PET-Membrane
Product Code. 16134957 Shop All Brand Products
Change view
Click to view available options
Pore Size:
0.4 μm
8 μm
Quantity:
4 x 12
48 x 1
Unit Size:
Pack of 48
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Quantity Unit Size
16134957 8 μm 4 x 12 Pack of 48
16104957 0.4 μm 48 x 1 Pack of 48
16114957 0.4 μm 4 x 12 Pack of 48
16124957 8 μm 48 x 1 Pack of 48
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16134957 Supplier BRAND™ Supplier No. 782717
Expédition estimée: 30-11-2026
Add to basket
Add to basket
This item is not returnable. Vea la política de devoluciones
The cellGrade™ plus surface of the membranes ensures excellent cell attachment and growth. The highly transparent membranes give you good visibilty of your cell cultures, and the high pore density contributes to good permeability.
  • Use Inserts in a hanging or standing position
  • For 24-well plates or culture dishes
  • Achieve uniform and controlled cell growth due to surface treatment, without coatings
TRUSTED_SUSTAINABILITY
Especificaciones
Description 24-well Insert 2in1, PET-Membrane 8 μm, Multipack
Material PET
Pore Size 8 μm
Quantity 4 x 12
Sterility Sterile
No. of Wells 24
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.