Learn More
Thermo Scientific Chemicals 4mm Die Set for KBr Quick Press Kit
Product Code. 15409189
Change view
Click to view available options
Sample Size:
2 mm
4 mm
7 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sample Size Unit Size
15409189 7 mm Each
15451557 4 mm Each
15461557 2 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15409189 Supplier Thermo Scientific Chemicals Supplier No. 039787.KT
 Restricted Item
Expédition estimée: 29-12-2026
Add to basket
This item is not returnable. View return policy
For use with KBr Quick Press Kit
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Die Set
Sample Size 7 mm
For Use With (Equipment) KBr Quick Press Kit