Learn More
3-(3,4-Dihydro-2H-pyrrol-5-yl)pyridine, 98%, BLD Pharm™
Product Code. 31002618
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
25 g
100 g
Unit Size:
100g
100mg
1g
250mg
25g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31002618 250 mg 250mg
30996891 5 g 5g
30993068 100 mg 100mg
30995062 25 g 25g
30999384 1 g 1g
30999924 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31002618 Supplier BLD Pharm Supplier No. BD140577250mg
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 532-12-7
Molecular Formula C9H10N2
MDL Number MFCD00052019
Quantity 250 mg
SMILES C1(C2=NCCC2)=CC=CN=C1
Molecular Weight (g/mol) 146.19
Percent Purity 98%
Chemical Name or Material 3-(3,4-Dihydro-2H-pyrrol-5-yl)pyridine