Learn More
Lenz Laborglasinstrumente™ 3.3 Borosilicate Glass Sedimentation Cone
Product Code. 10441093
Change view
Click to view available options
Capacity (Metric):
1000 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10441093 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10441093 Supplier Lenz Laborglasinstrumente™ Supplier No. 05670001
  • ONLINE EXCLUSIVE PRICE
    190.00€
    134.46€ / 1 pièces
    Économisez 55.54€
    29% de réduction
Expédition estimée: 16-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Capacity (Metric) 1000 mL
Material Borosilicate Glass (3.3)
Type Sedimentation Cone
Certifications/Compliance DIN 12672
Quantity 1 Pack