Learn More
3,3'-Iminodipropionitrile, TRC
Product Code. 30403089
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30403089 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30403089 Supplier Toronto Research Chemicals Supplier No. TRCI40901325G
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3,3'-Iminodipropionitrile
CAS 111-94-4
Molecular Formula C6H9N3
Quantity 25 g
Synonym 3,3'-Iminobis(propionitrile), BBCE, Bis(2-cyanoethyl)amine, Bis(beta-cyanoethyl)amine, Di(2-cyanoethyl)amine, 2-cyano-N-(2-cyanoethyl)-ethanamine, DPN, Iminobis(propionitrile), N,N-Bis(2-cyanoethyl)amine, NSC 7770, beta,beta'-Iminodipropionitrile
InChI Formula InChI=1S/C6H9N3/c7-3-1-5-9-6-2-4-8/h9H,1-2,5-6H2
SMILES N#CCCNCCC#N
IUPAC Name 3-(2-cyanoethylamino)propanenitrile
Molecular Weight (g/mol) 123.16
Formula Weight 123.0796