Learn More
3,4-Di(methoxy-d3)-benzoic Acid Methyl Ester, TRC
Product Code. 30379767
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30379767 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30379767 Supplier Toronto Research Chemicals Supplier No. TRCD463692100MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3,4-Di(methoxy-d3)-benzoic Acid Methyl Ester
CAS 1006717-12-9
Molecular Formula C10H6D6O4
Quantity 100 mg
InChI Formula InChI=1S/C10H12O4/c1-12-8-5-4-7(10(11)14-3)6-9(8)13-2/h4-6H,1-3H3/i1D3,2D3
SMILES [2H]C([2H])([2H])Oc1ccc(cc1OC([2H])([2H])[2H])C(=O)OC
IUPAC Name methyl 3,4-bis(trideuteriomethoxy)benzoate
Molecular Weight (g/mol) 202.24
Formula Weight 202.1112