Learn More
3,4-Dichlorobenzotrifluoride, TRC
Specifica
Specifica
| Chemical Name or Material | 3,4-Dichlorobenzotrifluoride |
| CAS | 328-84-7 |
| Molecular Formula | C7H3Cl2F3 |
| Quantity | 1 g |
| Synonym | 1,2-Dichloro-4-(trifluoromethyl)benzene, (3,4-Dichlorophenyl)trifluoromethane, 1-(Trifluoromethyl)-3,4-dichlorobenzene, 3,4-Dichloro(trifluoromethyl)benzene, 3,4-Dichloro-1-(trifluoromethyl)benzene, 3,4-Dichloro-alpha,alpha,alpha-trifluorotoluene, 4-(Trifluoromethyl)-1,2-dichlorobenzene, Oxsol 1000 |
| InChI Formula | InChI=1S/C7H3Cl2F3/c8-5-2-1-4(3-6(5)9)7(10,11)12/h1-3H |
| SMILES | FC(F)(F)c1ccc(Cl)c(Cl)c1 |
| IUPAC Name | 1,2-dichloro-4-(trifluoromethyl)benzene |
| Molecular Weight (g/mol) | 215 |
| Formula Weight | 213.9564 |