Learn More
3,4-Dihydroxybenzoic Acid Methyl Ester, TRC
Product Code. 30012593
Change view
Click to view available options
Quantity:
50 mg
1 g
Unit Size:
1g
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012593 50 mg 50mg
30006788 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012593 Supplier Toronto Research Chemicals Supplier No. TRCD45169550MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2150-43-8
Molecular Formula C8 H8 O4
Molecular Weight (g/mol) 168.15
Synonym Benzoic acid, 3,4-dihydroxy-, methyl ester, Protocatechuic acid, methyl ester (6CI,7CI,8CI), 3,4-Dihydroxybenzoic acid methyl ester, Methyl 3,4-dihydroxybenzoate, Methyl protocatechuate, NSC 146458
IUPAC Name methyl 3,4-dihydroxybenzoate
SMILES COC(=O)c1ccc(O)c(O)c1
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C8H8O4/c1-12-8(11)5-2-3-6(9)7(10)4-5/h2-4,9-10H,1H3
Formula Weight 168.0423
Chemical Name or Material 3,4-Dihydroxybenzoic Acid Methyl Ester