Learn More
3-(4-Methoxyphenyl)-3-oxopropanenitrile, 98%, BLD Pharm™
Product Code. 30987835
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
25 g
Unit Size:
10g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30987835 10 g 10g
30974745 250 mg -
30969325 1 g -
30971596 5 g -
30974986 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30987835 Supplier BLD Pharm Supplier No. BD1287110g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 3672-47-7
Molecular Formula C10H9NO2
MDL Number MFCD00067889
Quantity 10 g
SMILES COC1=CC=C(C=C1)C(=O)CC#N
Molecular Weight (g/mol) 175.18
Percent Purity 98%
Chemical Name or Material 3-(4-Methoxyphenyl)-3-oxopropanenitrile