missing translation for 'onlineSavingsMsg'
Learn More

3-Amidinopyridine hydrochloride, 97%

Product Code. 11444337
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444337 25 g 25g
11424337 1 g 1g
11434337 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444337 Supplier Thermo Scientific Alfa Aesar Supplier No. B20406.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7356-60-7 | C6H8ClN3 | 157.601 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7356-60-7
Molecular Formula C6H8ClN3
Molecular Weight (g/mol) 157.601
MDL Number MFCD00054326
InChI Key MKJPBOVLAZADQJ-UHFFFAOYSA-N
Synonym nicotinimidamide hydrochloride, 3-amidinopyridine hydrochloride, pyridine-3-carboximidamide hydrochloride, nicotinamidine hydrochloride, 3-pyridinecarboximidamide, monohydrochloride, nicotinamidine, hydrochloride, pyridine-3-carbonamidine hydrochloride, pyridine-3-carboxamidine hydrochloride, 3-pyridinecarboxamidine hydrochloride, pyridine-3-carboximidamide hcl
PubChem CID 202042
IUPAC Name pyridine-3-carboximidamide;hydrochloride
SMILES C1=CC(=CN=C1)C(=N)N.Cl

Specifications

Melting Point 187°C to 189°C
Quantity 25 g
Sensitivity Hygroscopic
Formula Weight 157.6
Percent Purity 97%
Chemical Name or Material 3-Amidinopyridine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.