Learn More
TPP Techno Plastic Products AG 3-B Cell Scraper S
Product Code. 17225336
Change view
Click to view available options
Length (Metric) Handle:
24 cm
Unit Size:
Pack of 40
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Handle Unit Size
17225336 24 cm Pack of 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17225336 Supplier TPP Techno Plastic Products AG Supplier No. 399002
  • ONLINE EXCLUSIVE PRICE
    320.00€
    245.31€ / 40 pièces
    Économisez 74.69€
    23% de réduction
En stock
Add to basket
Edge
Add to basket
3-B Cell scraper S, L = 24 cm, triple bagged
  • The triple bagged packaging (3-B) solution from TPP meets the increasingly more stringent clean room requirements.
  • The 3-B products provide noticeably more safety in grade clean aseptic room productions. With the standard packaging, contamination adhering to the outside of the bag could be unintentionally carried into cleanroom areas.
  • The 3-B packaging system prevents contamination from reaching highly sensitive production areas, thus significantly increasing safety for handling substances in a sterile environment.
  • All advantages of the standard product
  • Ideal for use in dedicated areas where the sterile transfer of product is essential
  • Each product is triple-wrapped in three peel-open bags
  • Each bag is marked with a security number: 3 – outer, 2 – middle, 1 – inner bag - the subsequent numbering helps to show status of packaging
  • Innermost bag 1 is labeled with the product data: product number and description, lot number and EXP.
  • The safety-open-style peel-open bags are increasing sized, so that during handling of one bag the next inner bag is not accidentally broken rendering it contaminated
Specifications
Length (Metric) Handle 24 cm
Product Type Triple-bagged