Learn More
(3-Bromo-1-propen-1-yl)benzene, 98%, BLD Pharm™
Product Code. 30968930
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30968930 25 g 25g
30968868 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30968930 Supplier BLD Pharm Supplier No. BD12136825g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 4392-24-9
Molecular Formula C9H9Br
MDL Number MFCD00000245
Quantity 25 g
SMILES BrCC=CC1=CC=CC=C1
Molecular Weight (g/mol) 197.07
Percent Purity 98%
Chemical Name or Material (3-Bromo-1-propen-1-yl)benzene