missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-(trifluoromethoxy)benzoyl chloride, 97%, Thermo Scientific™

Product Code. 15454407
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15454407 1 g 1g
15464407 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 15454407

Brand: Thermo Scientific Alfa Aesar H31775.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This product has been withdrawn. Where a replacement item has been identified it will be shown below
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1092461-36-3
Molecular Formula C8H3BrClF3O2
Molecular Weight (g/mol) 303.46
MDL Number MFCD11519379
InChI Key PRRWHYQCMQADHB-UHFFFAOYSA-N
Synonym 3-bromo-5-trifluoromethoxy benzoyl chloride, 3-bromo-5-trifluoromethoxy benzoyl chloride, jrd
PubChem CID 54772359
IUPAC Name 3-bromo-5-(trifluoromethoxy)benzoyl chloride
SMILES FC(F)(F)OC1=CC(Br)=CC(=C1)C(Cl)=O

Specifications

Refractive Index 1.506
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 303.46
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-(trifluoromethoxy)benzoyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.