Learn More
3-Chloro-1-(4-methoxyphenyl)propan-1-one, TRC
Product Code. 30360408
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30360408 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30360408 Supplier Toronto Research Chemicals Supplier No. TRCC596743500MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Chloro-1-(4-methoxyphenyl)propan-1-one
CAS 35999-20-3
Molecular Formula C10H11O2Cl
Quantity 500 mg
Synonym 3-Chloro-1-(4-methoxyphenyl)-1-propanone, 2-Chloroethyl 4-Methoxyphenyl Ketone, 3-Chloro-1-(4'-methoxyphenyl)propan-1-one, 3-Chloro-4'-methoxypropiophenone, NSC 31620, p-(3-Chloropropan-1-one)anisole, p-Methoxy-β-chloropropiophenone, β-Chloro-p-methoxypropiophenone
InChI Formula InChI=1S/C10H11ClO2/c1-13-9-4-2-8(3-5-9)10(12)6-7-11/h2-5H,6-7H2,1H3
SMILES COc1ccc(cc1)C(=O)CCCl
IUPAC Name 3-chloro-1-(4-methoxyphenyl)propan-1-one
Molecular Weight (g/mol) 198.65