Learn More
3-Chloro-1-(4-methoxyphenyl)propan-1-one, TRC
Product Code. 30359877
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30359877 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30359877 Supplier Toronto Research Chemicals Supplier No. TRCC5967431G
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Chloro-1-(4-methoxyphenyl)propan-1-one
CAS 35999-20-3
Molecular Formula C10H11O2Cl
Quantity 1 g
Synonym 3-Chloro-1-(4-methoxyphenyl)-1-propanone, 2-Chloroethyl 4-Methoxyphenyl Ketone, 3-Chloro-1-(4'-methoxyphenyl)propan-1-one, 3-Chloro-4'-methoxypropiophenone, NSC 31620, p-(3-Chloropropan-1-one)anisole, p-Methoxy-β-chloropropiophenone, β-Chloro-p-methoxypropiophenone
InChI Formula InChI=1S/C10H11ClO2/c1-13-9-4-2-8(3-5-9)10(12)6-7-11/h2-5H,6-7H2,1H3
SMILES COc1ccc(cc1)C(=O)CCCl
IUPAC Name 3-chloro-1-(4-methoxyphenyl)propan-1-one
Molecular Weight (g/mol) 198.65