Learn More
3-Chloro-5-methoxyaniline, TRC
Product Code. 30363374
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30363374 10 g
30360648 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30363374 Supplier Toronto Research Chemicals Supplier No. TRCC36790010G
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10272-06-7
Molecular Formula C7H8ClNO
Molecular Weight (g/mol) 157.6
Synonym 5-Chloro-m-anisidine, 3-Chloro-5-methoxybenzenamine
IUPAC Name 3-chloro-5-methoxyaniline
SMILES COc1cc(N)cc(Cl)c1
Specifications
Quantity 10 g
InChI Formula InChI=1S/C7H8ClNO/c1-10-7-3-5(8)2-6(9)4-7/h2-4H,9H2,1H3
Formula Weight 157.0294
Chemical Name or Material 3-Chloro-5-methoxyaniline