missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 15458855
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458855 250 mg 250mg
15448855 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15458855 Supplier Thermo Scientific Alfa Aesar Supplier No. H34258.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80194-68-9
Molecular Formula C7H3ClF3NO2
Molecular Weight (g/mol) 225.551
MDL Number MFCD00277482
InChI Key HXRMCZBDTDCCOP-UHFFFAOYSA-N
Synonym 3-chloro-5-trifluoromethyl pyridine-2-carboxylic acid, 3-chloro-5-trifluoromethyl picolinic acid, 2-pyridinecarboxylic acid, 3-chloro-5-trifluoromethyl, 3-chloro-5-trifluoromethyl-pyridine-2-carboxylic acid, 3-chloro-5-trifluoromethyl-2-pyridinecarboxylic acid, 2-carboxy-3-chloro-5-trifluoromethyl pyridine, ksc446c7n, 3-chloro-5-trifluoromethylpicolinic acid, 3-chloro-5-trifluoromethyl-picolinic acid
PubChem CID 2821908
IUPAC Name 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid
SMILES C1=C(C=NC(=C1Cl)C(=O)O)C(F)(F)F

Specifications

Melting Point 141°C to 143°C
Quantity 250 mg
Formula Weight 225.6
Percent Purity 97%
Chemical Name or Material 3-Chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.