Learn More
3-Chloropropiophenone, Mikromol™
Product Code. 30050921
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050921 25 mg 25mg
30051409 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050921 Supplier Mikromol Supplier No. MM0745.080025
 Restricted Item
Expédition estimée: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-Chloropropiophenone
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Chloropropiophenone
CAS 34841-35-5
Analyte or Component Names 3-Chloropropiophenone
Shipping Condition Room Temperature
Molecular Formula C9 H9 Cl O
Certifications/Compliance ISO 9001
Synonym 3-Chloropropane-1,2-diol, 3-Chloropropiophenone
InChI Formula InChI=1S/C9H9ClO/c1-2-9(11)7-4-3-5-8(10)6-7/h3-6H,2H2,1H3
SMILES CCC(=O)c1cccc(Cl)c1
IUPAC Name 1-(3-chlorophenyl)propan-1-one
Molecular Weight (g/mol) 168.6202
Quantity 25 mg
Formula Weight 168.0342 g/mol