missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-2-methyl-1-pentene, 99%, Thermo Scientific Chemicals

Product Code. 11459047
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11459047

Brand: Thermo Scientific Alfa Aesar B20624.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This product has been withdrawn. Where a replacement item has been identified it will be shown below
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19780-66-6
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00048664
InChI Key HPHHYSWOBXEIRG-UHFFFAOYSA-N
Synonym 3-ethyl-2-methyl-1-pentene, 1-pentene, 3-ethyl-2-methyl, 2-methyl-3-ethyl-1-pentene, acmc-1bqij, 1-pentene,3-ethyl-2-methyl
PubChem CID 140591
IUPAC Name 3-ethyl-2-methylpent-1-ene
SMILES CCC(CC)C(=C)C

Specifications

Density 0.72
Refractive Index 1.4124
Quantity 25 g
UN Number UN3295
Formula Weight 112.22
Percent Purity 99%
Chemical Name or Material 3-Ethyl-2-methyl-1-pentene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.