Learn More
3-Fluoro-4-(trifluoromethyl)aniline, 97%, BLD Pharm™
Product Code. 31431184
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
25 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31431184 1 g
31428783 10 g
31414623 25 g
31428330 250 mg
31425809 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31431184 Supplier BLD Pharm Supplier No. BD103691g
 Restricted Item
Expédition estimée: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 69411-68-3
Molecular Formula C7H5F4N
MDL Number MFCD00190120
Quantity 1 g
SMILES NC1=CC=C(C(F)(F)F)C(F)=C1
Molecular Weight (g/mol) 179.12
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-(trifluoromethyl)aniline