Learn More
Metrohm 3 mol/L KCI Electrolyte Solution
Product Code. 13253990
Encompass
Change view
Click to view available options
Filling Solution Type:
KCI Electrolyte Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Filling Solution Type
13253990 KCI Electrolyte Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13253990 Supplier Metrohm Supplier No. 6.2308.020
 Restricted Item
Expédition estimée: 22-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
An excellent solution for electrode conditioning.
  • Nonhazardous
  • For use with Ag/AgCl reference systems
  • Sold individually in a 250mL bottle
Specifications
For Use With (Equipment) Ag/AgCl Reference Systems
Filling Solution Type KCI Electrolyte Solution
Packaging 250 mL Bottle