missing translation for 'onlineSavingsMsg'
Learn More

3-Nitro-5-(trifluoromethyl)pyridin-2-ol, 97%, Thermo Scientific™

Product Code. 10307354
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10307354 25 g Amber Glass Bottle 25g
10374152 1 g Amber Glass Bottle 1g
10150842 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10307354 Supplier Thermo Scientific Maybridge Supplier No. RF04994EB
 Restricted Item
Expédition estimée: 27-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 33252-64-1
Molecular Formula C6H3F3N2O3
Molecular Weight (g/mol) 208.096
MDL Number MFCD00153193
InChI Key JYXKHKBZLLIWEV-UHFFFAOYSA-N
PubChem CID 2775093
IUPAC Name 3-nitro-5-(trifluoromethyl)-1H-pyridin-2-one
SMILES C1=C(C(=O)NC=C1C(F)(F)F)[N+](=O)[O-]

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 25 g
Synonym 2-hydroxy-3-nitro-5-trifluoromethyl pyridine, 3-nitro-5-trifluoromethyl pyridin-2 1h-one, 3-nitro-5-trifluoromethyl pyridin-2-ol, 3-nitro-5-trifluoromethyl-2-pyridinone, 2 1h-pyridinone,3-nitro-5-trifluoromethyl, 5-trifluoromethyl-3-nitropyridin-2-ol, 3-nitro-5-trifluoromethyl-1h-pyridin-2-one, 2 1h-pyridinone, 3-nitro-5-trifluoromethyl, 3-nitro-5-trifluoromethyl hydropyridin-2-one, pubchem9284
Formula Weight 208.09
Percent Purity 97%
Chemical Name or Material 3-Nitro-5-(trifluoromethyl)pyridin-2-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.