Learn More
3-Phenylfuran-2,5-dione, 97%, BLD Pharm™
Product Code. 30983557
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
Unit Size:
25g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30983557 10 g -
30973060 25 g 25g
30971975 1 g -
30969197 100 g -
30981060 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30983557 Supplier BLD Pharm Supplier No. BD1255710g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 36122-35-7
Molecular Formula C10H6O3
MDL Number MFCD00015472
Quantity 10 g
SMILES O=C(C(C1=CC=CC=C1)=C2)OC2=O
Molecular Weight (g/mol) 174.15
Percent Purity 97%
Chemical Name or Material 3-Phenylfuran-2,5-dione