Learn More
3-(Trifluoromethyl)benzoic acid, 98%, BLD Pharm™
Product Code. 30987628
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
1 kg
Unit Size:
10g
1kg
500g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30987628 10 g 10g
30982806 25 g -
30984068 100 g -
30987184 500 g 500g
30975754 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30987628 Supplier BLD Pharm Supplier No. BD1090210g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 454-92-2
Molecular Formula C8H5F3O2
MDL Number MFCD00002519
Quantity 10 g
SMILES O=C(O)C1=CC=CC(C(F)(F)F)=C1
Molecular Weight (g/mol) 190.12
Percent Purity 98%
Chemical Name or Material 3-(Trifluoromethyl)benzoic acid