Learn More
TubeWriter 300 - Custom Rack Side Print for Any Labware
Product Code. 30067433
Change view
Click to view available options
Product Type:
Labeling Rack
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
30067433 Labeling Rack Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30067433 Supplier TubeWriter Supplier No. PS0000XX300Custom
Expédition estimée: 18-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Custom racks made to exact specifications to print directly on side of unique labware
  • Custom racks designed for 300
  • Racks made to your exact specifications to directly print and label on the side of any labware
  • Rack will ensure correct alignment and holds labware securely
  • No time-consuming sticky labels or handwriting required
  • Racks designed for rapid switching between labware types
  • Print survives long-term storage at -80°C and liquid nitrogen
  • Print withstands the harshest lab conditions
  • Can print 2D or 3D barcodes, images and 5 lines of text
Specifications
Product Type Labeling Rack