missing translation for 'onlineSavingsMsg'
Learn More

DPP4/CD26 Mouse Monoclonal Antibody, Proteintech

Product Code. 30234657 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
Unit Size:
20µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30234657 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30234657 Supplier Proteintech Supplier No. 683831IG20UL
 Restricted Item
En stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The DPP4/CD26 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DPP4/CD26. This antibody recognizes human, pig, rabbit antigen. The DPP4/CD26 antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.

Specifications

Antigen DPP4/CD26
Applications Western Blot, Immunocytochemistry/Immunofluorescence, ELISA
Classification Monoclonal
Clone 2G6D11
Concentration 1000 μg/mL
Conjugate Unconjugated
Gene Symbols ADABP; ADCP2; CD26; DPPIV; TP103
Host Species Mouse
Immunogen Recombinant Protein
Purification Method Protein G purification
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1803
Target Species Human, Pig, Rabbit
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 110 kDa
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.