Learn More
3500/3500xL Data Collection Software, v3.3
Product Code. 18848728 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 license
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18848728 1 license Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18848728 Supplier Thermo Scientific Supplier No. A41857
 Restricted Item
Expédition estimée: 21-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
This product is a software upgrade package. It includes a 3500/3500xL Data Collection Software upgrade CD for upgrade from v3 or v3.1 to v3.3.
This product is a software upgrade package. It includes a 3500/3500xL Data Collection Software upgrade CD for upgrade from v3 or v3.1 to v3.3.
Data Collection Software v3.3 provides the features and functionality that researchers have been requesting to take their research even further. Powered by the Windows™ 10 operating system, our newest and most powerful data collection software provides:
  • Options to enable/disable consumable hard stop warnings
  • Option to enable/disable consumable hard stops for administrator
  • Signal Optimization Factor parameter added to spatial calibration
  • Bi-weekly maintenance option added to Scheduler
  • Ability to export consumable logs
  • Ability to export injection list
  • Ability to load another plate and re-order priorities after run is paused
  • Added run pause confirmation dialog
  • Support to load additional plate and re-order injections after run is paused
  • Ability to enable/disable off-scale recovery and pull-up reduction
  • Additional features include:
  • Support for Windows 10, 64 bit
  • Security—built-in SAE feature set assists in meeting more stringent requirements
  • Ability to upload sample files to Connect cloud platform in non-SAE mode
TRUSTED_SUSTAINABILITY
Specifications
Product Type Data Collection Software
Includes 3500/3500xL Data Collection Software v3/v3.3 to v3.3 CD, 1 licensing kit with single license code, combined RUO and SAE (Security, Audit, and E-signature), 1 user document CD
For Use With (Application) Data Collection
For Use With (Equipment) 3500 Genetic Analyzer
Operating System Microsoft Windows 10 IoT 64-bit
Quantity 1 license