Learn More
((3aR,4R,6R,6aR)-6-Methoxy-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxol-4-yl)methanol, 95%, BLD Pharm™
Spécification
Spécification
| CAS | 4099-85-8 |
| Molecular Formula | C9H16O5 |
| MDL Number | MFCD00270040 |
| Quantity | 100 g |
| SMILES | OC[C@@H](O1)[C@@H](O2)[C@@H](OC2(C)C)[C@@H]1OC |
| Molecular Weight (g/mol) | 204.22 |
| Percent Purity | 95% |
| Chemical Name or Material | ((3aR,4R,6R,6aR)-6-Methoxy-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxol-4-yl)methanol |