Learn More
Uvex™ 3B Chem Light Chemical Protection Coveralls
Product Code. 17737649 Shop All Uvex Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17737649 Small Pack of 25
17747649 Medium Pack of 25
17757649 Large Pack of 25
17767649 X-Large Pack of 25
17777649 2X-Large Pack of 25
17787649 3X-Large Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 17737649 Supplier Uvex™ Supplier No. 8984309
  • ONLINE EXCLUSIVE PRICE
    295.00€
    265.86€ / 25 pièces
    Économisez 29.14€
    9% de réduction
Expédition estimée: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Disposable coveralls constructed of polypropylene SMS laminated with polyethylene film for maximum safety.
  • Light and highly flexible material for high wearer comfort
  • Liquid-tight
  • Low-noise material
  • Free from paint wetting impairment substances and silicone
  • Ideal for cleaning work
  • Middle finger loops for securing the sleeves
  • Optimal protection thanks to self-adhesive zipper flap
  • Anti-static properties
  • Protection against infectious diseases
  • Lined with skin-friendly non-woven material
  • Secure and comfortable closures thanks to elasticated bands on hood, arms and legs
  • Elasticated waistband for optimal fit
  • Two-way zip
TRUSTED_SUSTAINABILITY
Specifications
Product Type Chemical Protective Clothing
Size Small
Gender Unisex
Material Polypropylene
Color White, Yellow
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Covered Elastic
Neck Style Hooded
Hood Type Standard
Closure Type Zipper with Self-adhesive Flap
Seam Type Taped
Sterility Non-sterile
Resistance Chemicals
Certifications/Compliance EN 14126, EN 14605 - type 3B, EN 14605 - type 4B, ISO 13982-1 - type 5B, EN 13034 - type 6B, EN 1149-5, EN 1073-2, ISO 27065 C3