Learn More
3M™ Versaflo™ Peel-Off Visor Cover for S-Series Hood Assembly
Product Code. 15789627 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
S-Series Hoods and Headcovers
Unit Size:
10 pairs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15789627 S-Series Hoods and Headcovers 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15789627 Supplier 3M™ Supplier No. 7000000156
  • ONLINE EXCLUSIVE PRICE
    50.00€
    34.27€ / 10 pièces
    Économisez 15.73€
    31% de réduction
Expédition estimée: 28-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Peel-off visor cover is for use with S-Series hoods and headcovers to protect from contaminants and scratches.
  • Transparent covering to hood
  • Protect from contaminants and scratches
  • 3M Catalog# S-922
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Disposable Reusable
Product Type Lens Cover
For Use With (Application) Protects Visor From Contaminants and Scratches
For Use With (Equipment) S-Series Hoods and Headcovers
Model S-922