Learn More
3M™ Versaflo™ Comfort Faceseal for M-100 and M-300 Series PAPR
Product Code. 13599030 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Versaflo™ M-100 and M-300 Series
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
13599030 Versaflo™ M-100 and M-300 Series Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13599030 Supplier 3M™ Supplier No. 7100137904
  • ONLINE EXCLUSIVE PRICE
    184.00€
    127.03€ / 5 pièces
    Économisez 56.97€
    30% de réduction
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Comfortable, low-linting faceseal for Versaflo™M-100 Series and M-300 Series products.
  • Low-linting faceseal
  • Comfortable
  • Color: Black
  • Headcover
  • 3M Catalog# M-936
TRUSTED_SUSTAINABILITY
Specifications
Disposable Reusable
Product Type Faceseal
For Use With (Application) Cleaning, Electrical, Facility Maintenance, Painting, Sanding, Welding
For Use With (Equipment) Versaflo™ M-100 and M-300 Series
Includes 4-point textile straps, visor
Model M-936