Learn More
3M™ Versaflo™ M-Series PAPR Flame Resistant Headgear
Product Code. 15760068 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
M-Series PAPR Systems
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15760068 M-Series PAPR Systems Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15760068 Supplier 3M™ Supplier No. 7100063882
  • ONLINE EXCLUSIVE PRICE
    55.00€
    37.41€ / 1 pièces
    Économisez 17.59€
    31% de réduction
Expédition estimée: 17-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Headgear cover protects against the buildup of particles on the outside surface of the headgear shell.
Made from flame resistant polyester
  • Disposable product to be discarded when damaged or soiled
  • 3M Catalog# M-972
TRUSTED_SUSTAINABILITY
Specifications
Disposable Reusable
Material Polyester
Product Type Headgear Cover
For Use With (Application) Cleaning, Electrical, Facility Maintenance, Painting, Sanding, Welding
For Use With (Equipment) M-Series PAPR Systems
Model M-972