Learn More
3M™ Versaflo M-Series PARP Visor Replacement Parts
Product Code. 15700088 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
M-series parp
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15700088 M-series parp Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15700088 Supplier 3M™ Supplier No. 7100064616
  • ONLINE EXCLUSIVE PRICE
    92.00€
    62.86€ / 1 pièces
    Économisez 29.14€
    31% de réduction
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The M-920 Visor Frame Assembly is a replacement part for 3M™ Versaflo M-Series products.
Includes two M-919 Visor Frame buttons.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Visor Frame Assembly
For Use With (Application) Cleaning, electrical, facility maintenance, painting, sanding, welding
For Use With (Equipment) M-series parp
Includes Two M-919 visor frame buttons