Learn More
4-(1,3-Dioxoisoindolin-2-yl)butanoic acid, 98%, BLD Pharm™
Product Code. 30962992
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
25 g
Unit Size:
100mg
1g
250mg
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30962992 1 g 1g
30961002 250 mg 250mg
30955411 5 g 5g
30962237 25 g 25g
30956783 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30962992 Supplier BLD Pharm Supplier No. BD138381g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 3130-75-4
Molecular Formula C12H11NO4
MDL Number MFCD00196079
Quantity 1 g
SMILES OC(=O)CCCN1C(=O)C2=C(C=CC=C2)C1=O
Molecular Weight (g/mol) 233.22
Percent Purity 98%
Chemical Name or Material 4-(1,3-Dioxoisoindolin-2-yl)butanoic acid