Learn More
4,4,4',4',5,5,5',5'-Octamethyl-2,2'-bi(1,3,2-dioxaborolane), 98%, BLD Pharm™
Product Code. 31432647
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31432647 500 g
31422593 100 g
31424749 1 kg
31426250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31432647 Supplier BLD Pharm Supplier No. BD14304500g
 Restricted Item
Expédition estimée: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 73183-34-3
Molecular Formula C12H24B2O4
MDL Number MFCD00799570
Quantity 500 g
SMILES CC1(C)C(C)(C)OB(B2OC(C)(C)C(C)(C)O2)O1
Molecular Weight (g/mol) 253.94
Percent Purity 98%
Chemical Name or Material 4,4,4',4',5,5,5',5'-Octamethyl-2,2'-bi(1,3,2-dioxaborolane)