missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Biphenylyl)-4-oxobutyric acid, 96%

Product Code. 15477038
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15477038 1 g 1g
15487038 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15477038 Supplier Thermo Scientific Alfa Aesar Supplier No. H60557.03
 Restricted Item
Expédition estimée: 09-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 36330-85-5 | C16H14O3 | 254.285 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36330-85-5
Molecular Formula C16H14O3
Molecular Weight (g/mol) 254.285
MDL Number MFCD00056701
InChI Key ZPAKPRAICRBAOD-UHFFFAOYSA-N
Synonym fenbufen, lederfen, cinopal, bufemid, napanol, 4-4-biphenylyl-4-oxobutyric acid, 3-4-phenylbenzoyl propionic acid, 3-4-biphenylylcarbonyl propionic acid, 4-biphenyl-4-yl-4-oxobutanoic acid, gamma-oxo 1,1'-biphenyl-4-butanoic acid
PubChem CID 3335
ChEBI CHEBI:31599
IUPAC Name 4-oxo-4-(4-phenylphenyl)butanoic acid
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)CCC(=O)O

Specifications

Melting Point 184°C to 187°C
Quantity 1 g
UN Number UN2811
Formula Weight 254.28
Percent Purity 96%
Chemical Name or Material 4-(4-Biphenylyl)-4-oxobutyric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.