Learn More
4,7-Dihydro-4-oxo-2-[(triphenylmethyl)amino]-3H-pyrrolo[2,3-d]pyrimidine-5-carbonitrile, TRC
Product Code. 30377796
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30377796 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30377796 Supplier Toronto Research Chemicals Supplier No. TRCD4534755MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4,7-Dihydro-4-oxo-2-[(triphenylmethyl)amino]-3H-pyrrolo[2,3-d]pyrimidine-5-carbonitrile
CAS 1019853-64-5
Molecular Formula C26H19N5O
Quantity 5 mg
InChI Formula InChI=1S/C26H19N5O/c27-16-18-17-28-23-22(18)24(32)30-25(29-23)31-26(19-10-4-1-5-11-19,20-12-6-2-7-13-20)21-14-8-3-9-15-21/h1-15,17H,(H3,28,29,30,31,32)
SMILES O=C1C2=C(NC=C2C#N)N=C(NC(C3=CC=CC=C3)(C4=CC=CC=C4)C5=CC=CC=C5)N1
IUPAC Name 4-oxo-2-(tritylamino)-4,7-dihydro-3H-pyrrolo[2,3-d]pyrimidine-5-carbonitrile
Molecular Weight (g/mol) 417.46
Formula Weight 417.16