Learn More
4-Acetylbenzenesulfonamide, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Acetylbenzenesulfonamide |
| CAS | 1565-17-9 |
| Molecular Formula | C8H9NO3S |
| Quantity | 1 g |
| Synonym | 4-(Aminosulfonyl)acetophenone, p-Acetobenzenesulfonamide, p-Acetylbenzenesulfonamide |
| InChI Formula | InChI=1S/C8H9NO3S/c1-6(10)7-2-4-8(5-3-7)13(9,11)12/h2-5H,1H3,(H2,9,11,12) |
| SMILES | CC(=O)c1ccc(cc1)S(=O)(=O)N |
| IUPAC Name | 4-acetylbenzenesulfonamide |
| Molecular Weight (g/mol) | 199.23 |
| Formula Weight | 199.0303 |