Learn More
4-(Boc-amino)-1-methylpyrrole-2-carboxylic Acid, TRC
Product Code. 30346241
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30346241 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30346241 Supplier Toronto Research Chemicals Supplier No. TRCB618585250MG
 Restricted Item
Expédition estimée: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-(Boc-amino)-1-methylpyrrole-2-carboxylic Acid
CAS 77716-11-1
Molecular Formula C11 H16 N2 O4
Quantity 250 mg
Synonym 4-tert-Butoxycarbonylamino-1-methyl-1H-pyrrole-2-carboxylic Acid, 4-[[(1,1-Dimethylethoxy)carbonyl]amino]-1-methyl-1H-pyrrole-2-carboxylic Acid
InChI Formula InChI=1S/C11H16N2O4/c1-11(2,3)17-10(16)12-7-5-8(9(14)15)13(4)6-7/h5-6H,1-4H3,(H,12,16)(H,14,15)
SMILES Cn1cc(NC(=O)OC(C)(C)C)cc1C(=O)O
IUPAC Name 1-methyl-4-[(2-methylpropan-2-yl)oxycarbonylamino]pyrrole-2-carboxylic acid
Molecular Weight (g/mol) 240.256
Formula Weight 240.111