Learn More
4-Bromoisophthalic acid, 98%, BLD Pharm™
Product Code. 31426751
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31426751 5 g
31422404 1 g
31426048 25 g
31425387 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31426751 Supplier BLD Pharm Supplier No. BD108735g
 Restricted Item
Expédition estimée: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 6939-93-1
Molecular Formula C8H5BrO4
MDL Number MFCD00002404
Quantity 5 g
SMILES OC(=O)C1=CC(C(O)=O)=C(Br)C=C1
Molecular Weight (g/mol) 245.03
Percent Purity 98%
Chemical Name or Material 4-Bromoisophthalic acid