Learn More
4-Ethoxy-3-(trifluoromethyl)benzylamine, 97%, Thermo Scientific™
Product Code. 15477247
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15477247 1 g 1g
15487247 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15477247 Supplier Thermo Scientific Alfa Aesar Supplier No. H34308.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1206593-27-2
Molecular Formula C10H12F3NO
Molecular Weight (g/mol) 219.21
MDL Number MFCD13196584
InChI Key GHCHXTNRZYWWPF-UHFFFAOYSA-N
Synonym 4-ethoxy-3-trifluoromethyl benzylamine, 1-4-ethoxy-3-trifluoromethyl phenyl methanamine, 4-ethoxy-3-trifluoromethyl phenyl methanamine, 4-ethoxy-3-trifluoromethyl phenyl methylamine
PubChem CID 57361411
IUPAC Name [4-ethoxy-3-(trifluoromethyl)phenyl]methanamine
SMILES CCOC1=C(C=C(CN)C=C1)C(F)(F)F
Specifications
Quantity 1 g
UN Number UN3259
Sensitivity Air sensitive
Formula Weight 219.21
Percent Purity 97%
Chemical Name or Material 4-Ethoxy-3-(trifluoromethyl)benzylamine