Learn More
4-Fluoroisobenzofuran-1,3-dione, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 652-39-1 |
| Molecular Formula | C8H3FO3 |
| MDL Number | MFCD00039696 |
| Quantity | 1 g |
| SMILES | O=C1OC(C2=C1C=CC=C2F)=O |
| Molecular Weight (g/mol) | 166.11 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-Fluoroisobenzofuran-1,3-dione |