Learn More
4-Methoxy-3-[3-(morpholin-4-yl)propoxy]benzaldehyde, TRC
Product Code. 30482496
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30482496 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30482496 Supplier Toronto Research Chemicals Supplier No. TRCM15214050MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Methoxy-3-[3-(morpholin-4-yl)propoxy]benzaldehyde
CAS 861453-11-4
Molecular Formula C15 H21 N O4
Quantity 50 mg
Synonym Benzaldehyde, 4-methoxy-3-[3-(4-morpholinyl)propoxy]-, 4-Methoxy-3-[3-(4-morpholinyl)propoxy]benzaldehyde, 4-Methoxy-3-[3-(morpholin-4-yl)propoxy]benzaldehyde
InChI Formula InChI=1S/C15H21NO4/c1-18-14-4-3-13(12-17)11-15(14)20-8-2-5-16-6-9-19-10-7-16/h3-4,11-12H,2,5-10H2,1H3
SMILES COc1ccc(C=O)cc1OCCCN2CCOCC2
IUPAC Name 4-methoxy-3-(3-morpholin-4-ylpropoxy)benzaldehyde
Molecular Weight (g/mol) 279.33
Formula Weight 279.1471