missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-(4-(trifluoromethyl)phenyl)-1,3-thiazole-5-sulfonyl chloride, 90%, Thermo Scientific™

Product Code. 10137362
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10137362 1 g 1g
10402593 250 mg 250mg
10275854 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10137362 Supplier Thermo Scientific Maybridge Supplier No. MO00860DA
 Restricted Item
Expédition estimée: 30-07-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 568577-83-3
Molecular Formula C11H7ClF3NO2S2
Molecular Weight (g/mol) 341.747
MDL Number MFCD04974054
InChI Key FIPBWMJINRBLKG-UHFFFAOYSA-N
Synonym 4-methyl-2-4-trifluoromethyl phenyl-1,3-thiazole-5-sulfonyl chloride, 4-methyl-2-4-trifluoromethyl phenyl thiazole-5-sulfonyl chloride
PubChem CID 2794632
IUPAC Name 4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-sulfonyl chloride
SMILES CC1=C(SC(=N1)C2=CC=C(C=C2)C(F)(F)F)S(=O)(=O)Cl

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 341.76
Percent Purity 90%
Chemical Name or Material 4-Methyl-2-[4-(trifluoromethyl)phenyl]-1, 3-thiazole-5-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.