Learn More
4'-n-Octylacetophenone, 98%, Thermo Scientific™
Product Code. 11494894
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494894 5 g 5g
11484894 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11494894 Supplier Thermo Scientific Alfa Aesar Supplier No. A13817.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10541-56-7
Molecular Formula C16H24O
Molecular Weight (g/mol) 232.37
MDL Number MFCD00043682
InChI Key GARQDIVXKVBJFP-UHFFFAOYSA-N
Synonym 1-4-octylphenyl ethanone, p-octylacetophenone, 4'-n-octylacetophenone, 4'-octylacetophenone, ethanone, 1-4-octylphenyl, p-n-octylacetophenone, 4-n-octylacetophenone, acetophenone, 4'-octyl, 1-acetyl-4-octylbenzene, p-octylaceto-phenone
PubChem CID 66344
IUPAC Name 1-(4-octylphenyl)ethanone
SMILES CCCCCCCCC1=CC=C(C=C1)C(C)=O
Specifications
Boiling Point 152°C to 153°C (1 mmHg)
Quantity 5 g
Formula Weight 232.37
Percent Purity 98%
Chemical Name or Material 4'-n-Octylacetophenone