Learn More
4-Nitro-3-(trifluoromethyl)aniline, Mikromol™
Product Code. 30457671
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457671 4 x 25 mg 100mg
30052648 100 mg 100mg
30049857 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457671 Supplier Mikromol Supplier No. MM0161.0300254AMP
 Restricted Item
Expédition estimée: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-Nitro-3-(trifluoromethyl)aniline, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-nitro-3-(trifluoromethyl)aniline
CAS 393-11-3
Analyte or Component Names 4-nitro-3-(trifluoromethyl)aniline
Shipping Condition Room Temperature
Molecular Formula C7 H5 F3 N2 O2
Certifications/Compliance ISO 17025
Synonym Flutamide Imp. A (EP), Flutamide USP Related Compound A, Flutamide USP RC A, 4-Nitro-3-(trifluoromethyl)aniline, Flutamide Related Compound A, Flutamide Impurity A
InChI Formula InChI=1S/C7H5F3N2O2/c8-7(9,10)5-3-4(11)1-2-6(5)12(13)14/h1-3H,11H2
Solution Type Neat
SMILES Nc1ccc(c(c1)C(F)(F)F)[N+](=O)[O-]
IUPAC Name 4-nitro-3-(trifluoromethyl)aniline
Molecular Weight (g/mol) 206.12
Quantity 4 x 25 mg
Formula Weight 206.0303 g/mol